Sanitary In-Line Strainer Assembly; 1.5" ; with 1/4" Perforated Core
Representative image only.
Item # EW-29590-12
Protect products and expensive process equipment from coarse or fine particulates
  • 316 SS fluid contact surfaces are polished to a smooth, blemish free finish
  • Standard clamp sizes allow easy installation or retrofit
PRODUCT OPTIONS
$1,128.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Filter TypeStrainer
  • Material316 Stainless Steel
  • Inlet Connection1-1/2" Tri-Clamp®
  • Outlet Connection1-1/2" Tri-Clamp®
  • Length (cm)40.01
  • Length (in)15 3/4
  • DescriptionSanitary In-Line Strainer Assembly; 1.5" ; with 1/4" Perforated Core
Includes
strainer assemblies include housing, perforated core, internal cap and spring, assembly clamp and gasket. Order wire mesh overscreens of the desired mesh size separately.
Key Features
  • 316 SS fluid contact surfaces are polished to a smooth, blemish free finish
  • Standard clamp sizes allow easy installation or retrofit
More About this Item
Sanitary strainers are ideal for particulate removal or pre-filtration in the processing of foods, beverages, pharmaceuticals, cosmetics, and other critical compounds. Housings are available in in-line and side-inlet configurations.