Weighing Unitsmg, g, kg, ct, lb, oz, ozt, Hong Kong taels, Singapore taels, Taiwanese taels, Chinese taels, GN, dwt, mom, tol, bat, MS, N
Calibration TypeInternal
OutputRS232, USB
USB InterfaceYes
RS-232 InterfaceYes
Stability IndicatorYes
Stabilization Time (Sec)2
Battery1 CR2032 Lithium (included)
Pan Length (in)3 19/64
Pan Width (in)3 19/64
Platform Length (in)15 29/32
Platform Width (in)9 2/5
Display Width (in)7
DescriptionCubis II Semimicro Balance 220g X 0.010mg with Pre-configured QApp 99 Software, Auto. Glass Draft shield & Integrated Ionizer
Warranty10 Years
Includes
120/230 VAC 50/60 Hz power adapter, glass draft shield, integrated ionizer, USB connection cable, dust cover, and CR2032 lithium battery .
Key Features
Fully customizable hardware, software, and connectivity
Complete data integrity for confident compliance
Error-free operation by assessing and correcting relevant factors
Advanced MCA user interface with swipe functionality
All-inclusive software package included
10 year warranty
More About this Item
Sartorius® Cubis® II MCA is a completely customizable balance to align with your unique application and compliance requirements while ensuring reliable results. Innovative technology features draft shield technology, fast and easy motorized leveling, an integrated ionizer for automatic electrostatic elimination, and climate sensor for environmental monitoring to ensure error-free operation.
Large 7” color touch screen display allows you to touch, scroll, and swipe with factory-installed weighing applications. The status center shows all information about your balance and environmental conditions as well as detailed help and support in case of warnings and errors.
The all-inclusive software, QApp 99, includes four different sub-packages for compliance, complex weighing applications, weighing applications and helpful tools, and connectors for data exchange. Full end-to-end data integrity follows ALCOA+ principles. Data cannot be manipulated with the user management function and LIMS connectivity. User management includes centralized password management support, high security management configuration, and login rules. Electronic signatures are in compliance with 21 CFR Part 11.
Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.