Sartorius QP10 Hardware Package Activation Code for Cubis® II Balance (MCA Models Only)
Representative image only.
Sartorius Mfr # QP10 – Item # EW-39743-45
Optimize performance of your Sartorius balance
  • Choose software packages based on your application needs
PRODUCT OPTIONS
$945.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithCubis® II MCA high-capacity micro balances 39743-15 to -24
  • DescriptionHardware Package Activation Code for Cubis® II MCA High-Capacity Micro Balance only
Key Features
  • Choose software packages based on your application needs
More About this Item
Add these software packages to activate lab applications, functions, or utilities on your Cubis balance. The licenses for these packages can be ordered together with the balance or later for installation after purchase. ( If ordering after your Cubis balances purchase, model and serial number is required to be supplied )
Activate both motorized draft shield and ionizer. Motorized draft shield provides touch-free operation for convenience and minimizes cross-contamination. The ionizer helps achieve a more stable and accurate reading by eliminating static.