Sartorius Tacta® LH-729070 Single-Channel, Adjustable-Volume Pipette; 100 to 1000 µL
Representative image only.
Sartorius Mfr # LH-729070 – Item # EW-24505-37
Superb comfort with effortless volume adjustment and locking
  • Achieve reliable results with the calibration adjustment for various fluid types
  • Hold comfortably with the ergonomically designed handle
  • Autoclave fully without disassembly for quick and easy cleaning
PRODUCT OPTIONS
$430.00USD / Each
$76.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Max Volume (µL)1000
  • AutoclavableYes
  • Min Volume (µL)100
  • Min Volume (mL)0.1
  • Max Volume (mL)1
  • Increments (µL)1
  • Accuracy (%)±2.0% to ±0.7%
  • Precision (%)±0.5% to ±0.2%
  • Number Of Channels1
  • Tip EjectorYes
  • Color CodeBlue
  • DescriptionSingle-Channel, Adjustable-Volume Pipette; 100 to 1000 µL
Two year warranty
Key Features
  • Achieve reliable results with the calibration adjustment for various fluid types
  • Hold comfortably with the ergonomically designed handle
  • Autoclave fully without disassembly for quick and easy cleaning
More About this Item
This premium mechanical pipette has the Optiload feature in both single-, and multichannel models for light tip loading with perfect tip sealing. The Optiject soft-tip ejection feature detaches the tip in a smooth, controlled manner. Optilock’s dual-function volume lock offers protection against accidental volume changes.
Use the easy-to-read, four-digit volume display to set exact volumes. Different volume options are color-coded for quick selection of corresponding pipette tips. Safe-Cone filters (included in models above 10 μL) protect against contamination. These pipettors are made of materials which have high chemical and UV resistance to ensure a long life span.