Sartorius Tacta® LH-729140 Eight-Channel, Adjustable-Volume Pipette; 30 to 300 µL
Representative image only.
Sartorius Mfr # LH-729140 – Item # EW-24505-43
Superb comfort with effortless volume adjustment and locking
  • Achieve reliable results with easy calibration adjustment for various fluid types
  • Hold comfortably with the ergonomically designed handle
  • Autoclave fully without disassembly for quick and easy cleaning
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Max Volume (µL)300
  • AutoclavableYes
  • Min Volume (µL)30
  • Min Volume (mL)0.03
  • Max Volume (mL)0.3
  • Increments (µL)0.2
  • Accuracy (%)±2.0% to ±0.6%
  • Precision (%)±1.0% to ±0.25%
  • Number Of Channels8
  • Tip EjectorYes
  • Color CodeOrange
  • DescriptionEight-Channel, Adjustable-Volume Pipette; 30 to 300 µL
  • Warranty2 year
Two year warrantyConformite Europeenne
Key Features
  • Achieve reliable results with easy calibration adjustment for various fluid types
  • Hold comfortably with the ergonomically designed handle
  • Autoclave fully without disassembly for quick and easy cleaning
  • 2 year warranty
More About this Item
This premium mechanical pipette has the Optiload feature in both single- and multichannel models for light tip loading with perfect tip sealing. The Optiject soft-tip ejection feature detaches the tip in a smooth, controlled manner. Optilock’s dual-function volume lock offers protection against accidental volume changes.
Use the easy-to-read, four-digit volume display to set exact volumes. Different volume options are color-coded for quick selection of corresponding pipette tips. Safe-Cone filters (included in models above 10 μL) protect against contamination. Pipettors are made of materials which have high chemical and UV resistance to ensure a long life span.