Sartorius YSH30 Filter Holder, 150 mm

Representative image only.
Sartorius Mfr # YSH30 – Item # EW-39742-66
Optimize performance of your Cubis II balance
  • Allows easy weighing of filters up to a diameter of 150 mm
  • Compatible with semi-micro and analytical balances

PRODUCT OPTIONS

$1,311.20USD / Each
$1,490.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Accessory TypeFilter holder
  • Diameter (in)150
  • DescriptionFilter Holder, 150 mm

Key Features

  • Allows easy weighing of filters up to a diameter of 150 mm
  • Compatible with semi-micro and analytical balances

More About this Item

Replacement parts and accessories for your Sartorius Cubis II series balances.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat