This item is now discontinued. We recommend the following as suggested replacements: EW-06101-40

Savillex Wide-Mouth Sample Bottle, PFA; 120 mL

Representative image only.
Savillex Mfr # 100-0120-01 – Item # EW-06103-00
Safely store your samples in chemically inert PFA jars
  • Constructed of durable and chemically inert Perfluoroalkoxy (PFA)
  • Comfortable grip and effortless handling with four ribbed areas
  • Easily view samples with translucent jar
  • Jar features 120 mL (4 oz) capacity
  • 3 month warranty

SPECS & DESCRIPTION

Specifications & Description

  • Capacity (mL)120
  • Capacity (oz)4
  • MaterialPerfluoroalkoxy (PFA)
  • Cap / Stopper Size70 mm (not included)
  • Bottle ColorTranslucent
  • AutoclavableYes
  • Withstands High Temperature (° F)500
  • Withstands Low Temperature (° F)-328
  • Outside Diameter (OD) (in)2 39/64
  • Outside Diameter (OD) (cm)6.63
  • Overall Height (in)2 5/32
  • Overall Height (cm)5.46
  • DescriptionWide-Mouth Sample Bottle, PFA; 120 mL
  • Warranty3 month
3 Month Warranty

Key Features

  • Constructed of durable and chemically inert Perfluoroalkoxy (PFA)
  • Comfortable grip and effortless handling with four ribbed areas
  • Easily view samples with translucent jar
  • Jar features 120 mL (4 oz) capacity
  • 3 month warranty

More About this Item

Seamless wide-mouth jars handle severe corrosives, solvents, sensitive reagents, or staining solutions. Withstand temperatures from ‑328°F to 500°F (‑200°C to 260°C). Knurled closures feature an inner secondary seal for permanent storage.

Jar accepts 70 mm cap. Cap not included, order separately.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat