Schott Pipe Tee, Schedule 80, PVC, 1in. NPT(F)
Representative image only.
Schott Mfr # 3405-010 – Item # EW-05994-61
Change electrodes quickly with a single twist
$10.80USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionPipe Tee, Schedule 80, PVC, 1in. NPT(F)
More About this Item
ERP and Viton® O-rings seal 12-mm electrode in housing for waterproof protection. Two stainless-steel pins lock the PVDF housing into a collar. The collar can be permanently mounted in the Schott pipe tee.