Scienceware 11646-0614 Safety Labeled Wash Bottle, soap

Representative image only.
Scienceware Mfr # 11646-0614 – Item # EW-06256-11

$43.25USD / of 6

BUY MORE & SAVE

  • 1-5$43.25USD / of 6
  • 6-29$38.93USD / of 6
  • 30-119$36.76USD / of 6
  • 120+$34.60USD / of 6
Often bought with…

SPECS & DESCRIPTION

Specifications & Description

  • Capacity (mL)500
  • Capacity (oz)16
  • MaterialLDPE
  • Cap MaterialPolyallomer (PA)
  • Cap / Stopper Size43 mm
  • Cap ColorBlue
  • AutoclavableNo
  • Label TypeSoap
  • Outside Diameter (OD) (in)2
  • Outside Diameter (OD) (cm)7.3
  • Overall Height (in)8 3/4
  • Overall Height (cm)22.5
  • DescriptionSafety Labeled Wash Bottle, soap

More About this Item

Bottles include a leakproof 43-mm polyallomer (PA) cap and nozzle assembly. Large lettering and color-coded caps allow quick identification of contents.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat