Scienceware 18513-2040 Serum Vial Rack for 16 to 20-mm Vials

Representative image only.
Scienceware Mfr # 18513-2040 – Item # EW-06732-20
Withstands elevated water temperatures for incubation
  • End plate handles
  • Steam autoclavable at 121 ºC (250 ºF)

SPECS & DESCRIPTION

Specifications & Description

  • Min Tube Size (mm)16
  • Max Tube Size (mm)20
  • Tubes Held40
  • Length (in)10
  • Width (in)4 3/8
  • Height (in)3 1/2
  • Length (mm)254
  • Width (mm)111.125
  • MaterialPolypropylene(PP)
  • DescriptionSerum vial rack for 16- to 20-mm vials

Key Features

  • End plate handles
  • Steam autoclavable at 121 ºC (250 ºF)

More About this Item

These polypropylene racks fit most water baths for elevated temperature incubations. They feature long-lasting construction and are corrosion resistant. Height from base to shelf is 1-5/8" (41 mm). Use these racks with 10-13 mm serum vials, RIA vials, and T3-T4 reaction vials (13-16 mm).

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat