Scienceware 19570 Barbed fittings, Straight/reducing connector pack
Representative image only.
Scienceware Mfr # 19570 – Item # EW-06460-00
Bundled in assortment packs for convenience and to save you money!
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
PRODUCT OPTIONS
$64.00USD / Pkg of 22
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPolypropylene
  • Connection TypeBarb
  • DescriptionBarbed fittings, Straight/reducing connector pack
Key Features
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
More About this Item
Barbed fittings are designed for quick and easy assembly without tools. Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi). Just slip your flexible tubing or hose onto the barb to create a secure seal.
This kit has two of each of the following fittings (in inches) ...
For straight connections:
1/8
3/16
1/4
3/8
1/2
For reducing connectors:
1/4 x 3/16
3/8 x 3/16
1/2 x 3/16
3/8 x 1/4
1/2 x 1/4
3/8 x 1/2