Scienceware 20307-1825 Therm-O-Vac Adapter of PTFE, size 24/40
Representative image only.
Scienceware Mfr # 20307-1825 – Item # EW-06140-27
$164.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Taper Size24/40
  • MaterialPolytetrafluoroethylene (PTFE)
  • DescriptionTherm-O-Vac Adapter of PTFE, size 24/40
More About this Item
For vacuum work or inlet/outlet of gases. Port accepts thermometer, glass tubing, or pipettes. Hose barb connection is of PTFE and accepts 5/16" tubing.