Scienceware 20309-1675 Thermometer Adapter of PTFE, size 10/18
Representative image only.
Scienceware Mfr # 20309-1675 – Item # EW-06140-12
$80.30USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Taper Size10/18
  • Port Size (ID) (mm)6 to 7
  • MaterialPolytetrafluoroethylene (PTFE)
  • DescriptionThermometer Adapter of PTFE, size 10/18
More About this Item
For air-tight vacuum work. Threaded PP screw cap on top with an inside O-ring for fine depth adjustment of thermometers or tubing.