Scienceware 20311-1717 Tubing Adapter of PTFE, size 24/40, 8 mm
Representative image only.
Scienceware Mfr # 20311-1717 – Item # EW-06140-52
$69.90USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Taper Size24/40
  • Port Size (ID) (mm)8
  • MaterialPolytetrafluoroethylene (PTFE)
  • DescriptionTubing Adapter of PTFE, size 24/40, 8 mm
More About this Item
For vacuum work or inlet/outlet of gases. Port accepts thermometer, glass tubing, or pipettes. Hose barb connection is of PTFE and accepts 5/16" tubing.