Scienceware F36931-0001 Earth-Friendly 1001 Stick, 500 case
Representative image only.
Scienceware Mfr # F36931-0001 – Item # EW-06263-86
Lessen environmental impact
  • FDA compliant sticks are safe for food, drug, cosmetic contact
  • Green alternative to petroleum-based sampling tools
PRODUCT OPTIONS
$261.50USD / Case of 500
SPECS & DESCRIPTION
Specifications & Description
  • Width (in)3/4
  • Width (cm)1.91
  • MaterialPolylactic Acid (PLA)
  • SterileNo
  • DescriptionEarth-Friendly 1001 Stick, 500 case
Key Features
  • FDA compliant sticks are safe for food, drug, cosmetic contact
  • Green alternative to petroleum-based sampling tools
More About this Item
These earth-friendly sampling sticks are durably constructed with 100% PLA (polylactic acid) resin. Whether you are mixing samples, handling capsules, scraping containers, or positioning solid samples for examination, these sampling sticks are up to the task.
PLA is a 100% plant-based material which contributes less greenhouse gasses from source to resin than its petroleum-based counterparts. Using annually renewable resources with little environmental impact, these sticks offer a low carbon footprint, are biodegradable, and can be disposed of through industrial composting, incineration, land fill, or recycling. They are comparable in strength and durability to polystyrene, polypropylene, and other plastic resin sampling tools. Nonsterile.