Scienceware F371220040 Polygon Spinbar, Magnetic Stir Bar w/Ring, 40 x 8 mm; 1/Pk
Representative image only.
Scienceware Mfr # F371220040 – Item # EW-04775-13
$8.10USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Diameter (in)0.315
  • Diameter (cm)0.8
  • DescriptionPolygon Spinbar, Magnetic Stir Bar w/Ring, 40 x 8 mm; 1/Pk
More About this Item
Center molded pivot ring minimizes the contact area between the bar and vessel; reducing friction and chattering.