Scienceware F371220060 Polygon Spinbar, Magnetic Stir Bar w/Ring, 60 x 10 mm; 1/Pk
Representative image only.
Scienceware Mfr # F371220060 – Item # EW-04775-17
$12.10USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Diameter (in)0.394
  • Diameter (cm)1
  • DescriptionPolygon Spinbar, Magnetic Stir Bar w/Ring, 60 x 10 mm; 1/Pk
More About this Item
Center molded pivot ring minimizes the contact area between the bar and vessel; reducing friction and chattering.