Scientific Industries Genie SI-1150 Roto-Shake 3-D Orbital Shaker Attachment
Representative image only.
Scientific Industries Mfr # SI-1150 – Item # EW-51603-84
Expand the capabilities of your shaker
  • 3-D orbital attachment for Roto-Shake
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithRoto-Shake Genie
  • DescriptionRoto-Shake 3-D Orbital Shaker Attachment
Key Features
  • 3-D orbital attachment for Roto-Shake
More About this Item
The 3-D Orbital Shaker Attachment provides gentle yet thorough mixing of dishes, flasks, beakers and open containers. It moves at a 5 degree angle through a 360 degree rotation (providing a combination of vertical and horizontal motions). This means that every corner or crevice of a container will receive complete mixing, making it the perfect choice for applications such as gel and blot staining/blocking, sample washing, polymer resin preparation, and more.