Scientific Industries Mini-300 Orbital-Genie Variable Speed Orbital Shaker, universal clamps, 230 VAC (EU plug)
Representative image only.
Scientific Industries Mfr # SI-M1502 – Item # EW-51820-08
Compact, durable, and user friendly
  • Small footprint takes up less bench space
  • Variable speed provides flexibility
  • Analog control is easy to use
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionOrbital-Genie Variable Speed Orbital Shaker, universal clamps, 230 VAC (EU plug)
  • Warranty2 year
Two year warranty
Key Features
  • Small footprint takes up less bench space
  • Variable speed provides flexibility
  • Analog control is easy to use
  • 2 year warranty
More About this Item
These space-saving benchtop orbital shakers offer variable speeds up to 300 rpm, perfect for applications ranging from gentle mixing to vigorous agitation. Analog dial lets you adjust speed to optimize mixing. A wide variety of platform options ensures accommodation of your sample containers, whether they are flasks, bottles, beakers, tubes, or dishes.