Scientific Industries SI-4100 MultiPlate Genie® High-Speed Vortex Microplate Shaker; 120V

Representative image only.
Scientific Industries Mfr # SI-4100 – Item # EW-51402-31
Provides uniform mixing in each microplate well
  • Handles up to 4 microplates
  • Compact and rugged design
  • Fully programmable
  • No need for calibration
  • Ideal of use in cold rooms or incubators

PRODUCT OPTIONS

$995.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Orbit Size (mm)1
  • Min Speed (rpm)500
  • Max Speed (rpm)3000
  • Max Load (lbs)1.1
  • Max Load (kg)0.5
  • Capacity4 microplates
  • Width (in)10
  • Height (in)6
  • Depth (in)15
  • Width (cm)25
  • Height (cm)15
  • Depth (cm)38
  • Timer Range0 to 99 minutes or continuos
  • Power (VAC)120
  • DescriptionMultiPlate Genie® High-Speed Vortex Microplate Shaker; 120V, North America Plug
  • Warranty2 years parts & labor
Made in the U.S.A.2 Year Warranty

Includes

Microplate tray and four microplates.

Key Features

  • Handles up to 4 microplates
  • Compact and rugged design
  • Fully programmable
  • No need for calibration
  • Ideal of use in cold rooms or incubators
  • Fast and easy loading and unloading of plates saves time
  • 2 years part and labor warranty

More About this Item

The high-speed vortex microplate shaker offers consistent well-to-well mixing in up to 4 microplates, (handles up to 8 microplates with optional accessory tray). Works with PCR plates, deep well plates, 96-well microplates, and 384-well microplates.

The fully programable unit allows you to adjust the time and speed ideal for your microplate sample mixing applications. Easily set the program for timed, continuous run, or interval mixing. For interval mixing, press the pulse feature on the keypad and set the time on and time off. The combined 1 mm orbit and speed setting provides accurate and reliable results every time.

Ideal for many microplate mixing applications such as PCR preparation, digestions, bead mixing, ELISA, EIA, and colorimetric assay.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat