SciMatCo SC9040 Flammable Storage Cabinet

Representative image only.
SciMatCo Mfr # SC9040 – Item # EW-86434-02
Compact size is perfect for laboratory benchtop chemical storage
  • 1 year warranty

PRODUCT OPTIONS

$958.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • MaterialPlywood
  • Capacity (Liters)30.3
  • Capacity (Gal)8
  • Width (in)31
  • Height (in)17
  • Depth (in)14 1/2
  • Width (cm)78.7
  • Height (cm)43.2
  • Depth (cm)36.8
  • ColorYellow
  • ComplianceOSHA, NFPA (flammable cabinet)
  • Shipping Weight (lbs)60
  • DescriptionFlammable Storage Cabinet, UL Listed
  • Warranty1 Year
1 Year Warranty

Key Features

  • 1 year warranty

More About this Item

Rugged cabinets are constructed of 1” (2.5 cm) thick, high-density plywood and finished with clear epoxy paint for superior durability. Liquid-tight, 2” (5.1 cm) trough at base of cabinet contains any spillage. Color and highly visible 8” (20.3 cm) hazard label ensures easy identification of cabinet type. These versatile cabinets are designed for the laboratory bench but also can be stacked three high, using optional floor stand, for a complete chemical storage center.


Compliance: OSHA, NFPA

Approvals:UL Listed

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat