Secondary Formaldehyde Filters for 42" Self-Monitoring Ductless Fume Hoods
Representative image only.
Mfr # AU-5507 – Item # EW-33713-76
SPECS & DESCRIPTION
Specifications & Description
  • Hood Width (in)42
  • Exterior Width (in)42
  • Filter TypeSecondary Formaldehyde
  • DescriptionSecondary Formaldehyde Filters for 42\" Self-Monitoring Ductless Fume Hoods
More About this Item
Protect yourself from hazardous fumes, vapors, odors, particles, and contaminants with one of four types of filters for 42” self-monitoring ductless fume hoods
The secondary formaldehyde filter is ideal for formaldehyde and glutaraldehyde.