Secondary Hydrocarbon Filter for 42" Self-Monitoring Ductless Fume Hood (2 required)
Representative image only.
Mfr # AU-5504 – Item # EW-33713-56
SPECS & DESCRIPTION
Specifications & Description
  • Hood Width (in)42
  • Exterior Width (in)42
  • Filter TypeSecondary Hydrocarbon
  • DescriptionSecondary Hydrocarbon Filter for 42\" Self-Monitoring Ductless Fume Hood (2 required)
More About this Item
Protect yourself from hazardous fumes, vapors, odors, particles, and contaminants with one of four types of filters for 42” self-monitoring ductless fume hoods
Secondary filter protects from dust particles, smoke, acid mists, and other submicron particles. The activated hydrocarbon filter provides a high degree of absorbency—ideal for organic vapors, odors and fumes, including alcohols, acetone, xylene, and methylene chloride.