Self-Monitoring Ductless Fume Hood, 42", 115 VAC

Representative image only.
Mfr # AU-550E – Item # EW-33713-10
  • Airflow display lets you know at a glance that you're working in a clean-air environment
  • Audible and visual alarms signal you to replace the filters
  • Exceeds OSHA and ANSI standard Z9.5

$11,815.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Hood Width (in)42
  • Blower TypeStandard
  • LightsFluorescent
  • Exterior Width (in)42
  • Exterior Height (in)48
  • Exterior Depth (in)28
  • Interior Width (in)42
  • Power (VAC)115
  • Power (Hz)60
  • DescriptionSelf-Monitoring Ductless Fume Hood, 42", 115 VAC
Two year warranty

Key Features

  • Airflow display lets you know at a glance that you're working in a clean-air environment
  • Audible and visual alarms signal you to replace the filters
  • Exceeds OSHA and ANSI standard Z9.5

More About this Item

Get full protection from hazardous fumes, vapors, odors, particles, and contaminants with these self-monitoring ductless fume hoods. The advanced monitoring and filter system act with synergy to ensure a safe working environment. This advanced monitoring system continuously senses both hydrocarbon output and air velocity levels. The LED screen displays airflow, filter type and application, and filter saturation level and lets you know immediately if you are working in safe conditions. Audible and visual alarms alert you to reduced airflow, filter condition, blower failure, and maximum time allowance.
The filter system incorporates a prefilter, a primary filter, and a secondary safety filter to provide added protection. The prefilter(s) protects the primary and secondary filters from dust particles, smoke, acid mists, and other submicron particles. Choose from four filter types in the tables below. The activated hydrocarbon filter provides a high degree of adsorbency<emdash>ideal for organic vapors, odors and fumes, including alcohols, acetone, xylene, and methylene chloride. Choose the mineral acid filter for hydrochloric and nitric acids and the formaldehyde filter for formaldehyde and glutaraldehyde. The HEPA filter has an efficiency rating of 99.997% for collecting particles down to 0.3 microns<emdash>use for containment of particles such as toxic powders, asbestos, and lead. Hoods measure 48"H x 28"D.
What's included: prefilters, two 17-watt fluorescent lights, and a deep chemical-resistant fiberglass spill tray.

ACCESSORIES

Accessories

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat