Sensidyne 106S Gas Detection Tubes, Carbon monoxide (SEI)
Representative image only.
Sensidyne Mfr # 106S – Item # EW-81900-74
Measure a wide variety of gases and vapors with this detection system
  • Hundreds of direct-read tubes available to meet your changing needs
PRODUCT OPTIONS
$127.00USD / Box of 10
SPECS & DESCRIPTION
Specifications & Description
  • Gas MeasuredCarbon monoxide (SEI)
  • Range10 to 250 ppm
  • TLV (ppm)25
  • DescriptionGas Detection Tubes, Carbon monoxide (SEI)
Key Features
  • Hundreds of direct-read tubes available to meet your changing needs
More About this Item
Stable absorbing media ensure distinct color changes for clear, accurate results. Calibration scales are printed on the basis of individual production lot, eliminating variation in tube diameter, packing precision, and reactivity.