Sensidyne 109U Gas Detection Tube, Chlorine, 10/bx

Representative image only.
Sensidyne Mfr # 109U – Item # EW-81901-31
Measure a wide variety of gases and vapors with this detection system
  • Hundreds of direct-read tubes available to meet your changing needs

PRODUCT OPTIONS

$103.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Gas MeasuredChlorine
  • Range0.05 to 2 ppm
  • DescriptionGas Detection Tube, Chlorine, 10/bx

Key Features

  • Hundreds of direct-read tubes available to meet your changing needs

More About this Item

Stable absorbing media ensure distinct color changes for clear, accurate results. Calibration scales are printed on the basis of individual production lot, eliminating variation in tube diameter, packing precision, and reactivity.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat