Sensidyne 158S Gas Detection Tube, Styrene, 10/bx
Representative image only.
Sensidyne Mfr # 158S – Item # EW-81901-44
Measure a wide variety of gases and vapors with this detection system
  • Hundreds of direct-read tubes available to meet your changing needs
  • 2 year warranty
PRODUCT OPTIONS
$111.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Gas MeasuredStyrene
  • Range2.5 to 300 ppm
  • DescriptionGas Detection Tube, Styrene, 10/bx
  • Warranty2 year
Two year warranty
Key Features
  • Hundreds of direct-read tubes available to meet your changing needs
  • 2 year warranty
More About this Item
Stable absorbing media ensure distinct color changes for clear, accurate results. Calibration scales are printed on the basis of individual production lot, eliminating variation in tube diameter, packing precision, and reactivity.