Sensidyne 159SC Gas Detection Tubes, Oxygen (Twin Tubes (5 Tests/Box))
Representative image only.
Sensidyne Mfr # 159SC – Item # EW-81900-32
Measure a wide variety of gases and vapors with this detection system
  • Hundreds of direct-read tubes available to meet your changing needs
  • 2 year warranty
PRODUCT OPTIONS
$148.25USD / Box of 10
SPECS & DESCRIPTION
Specifications & Description
  • Gas MeasuredOxygen
  • Range1.5 to 24%
  • DescriptionGas Detection Tubes, Oxygen (Twin Tubes (5 Tests/Box))
  • Warranty2 year
Two year warranty
Key Features
  • Hundreds of direct-read tubes available to meet your changing needs
  • 2 year warranty
More About this Item
Stable absorbing media ensure distinct color changes for clear, accurate results. Calibration scales are printed on the basis of individual production lot, eliminating variation in tube diameter, packing precision, and reactivity.