Sensidyne 173SA Gas Detection Tubes, Hydrogen Chloride
Representative image only.
Sensidyne Mfr # 173SA – Item # EW-81901-47
Measure a wide variety of gases and vapors with this detection system
  • Hundreds of direct-read tubes available to meet your changing needs
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Gas MeasuredHydrogen chloride
  • Range20 to 1200 ppm
  • DescriptionGas Detection Tubes, Hydrogen Chloride
  • Warranty1 year
1 Year Warranty
Key Features
  • Hundreds of direct-read tubes available to meet your changing needs
  • 1 year warranty
More About this Item
Stable absorbing media ensure distinct color changes for clear, accurate results. Calibration scales are printed on the basis of individual production lot, eliminating variation in tube diameter, packing precision, and reactivity.