Sensidyne 8014-120SD Gas Detection Tubes, Hydrogen sulfide (SEI)
Representative image only.
Sensidyne Mfr # 8014-120SD – Item # EW-81900-84
Measure a wide variety of gases and vapors with this detection system
  • Hundreds of direct-read tubes available to meet your changing needs
PRODUCT OPTIONS
$141.25USD / Box of 10
SPECS & DESCRIPTION
Specifications & Description
  • Gas MeasuredHydrogen sulfide (SEI)
  • Range1 to 60 ppm
  • TLV (ppm)10
  • DescriptionGas Detection Tubes, Hydrogen sulfide (SEI)
Key Features
  • Hundreds of direct-read tubes available to meet your changing needs
More About this Item
Stable absorbing media ensure distinct color changes for clear, accurate results. Calibration scales are printed on the basis of individual production lot, eliminating variation in tube diameter, packing precision, and reactivity.