Sensorex BV1 Ball Valve; Full-Port, CPVC, 1" NPT(F)
Representative image only.
Sensorex Mfr # BV-1 – Item # EW-27015-80
Easily remove and clean sensors without shutting system down
  • Remove sensors for cleaning and maintenance without system shutdown
$94.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Port Size (NPT(F))1
  • Max Pressure (PSI)100
  • Max Temperature (° C)80
  • Wetted MaterialsCPVC
  • DescriptionBall Valve; Full-Port, CPVC, 1" NPT(F)
Key Features
  • Remove sensors for cleaning and maintenance without system shutdown
More About this Item
The full port 1” ball valve mounts a pH sensor in pressurized lines or the sidewall of tanks. Sensors can be retracted for service without shutting down lines or the need to empty tanks. Constructed of durable CPVC. The valve is compatible with Sensorex insertion assemblies and sensors.