Sensorex CX2000 Contacting Conductivity Transmitter/Controller, 1/4 DIN
Representative image only.
Sensorex Mfr # CX2000 – Item # EW-19066-02
Contacting conductivity measurements from ultrapure to high conductivity measurements
  • Large, graphical LCD with auto-sense backlight and contrast can be easily viewed in all lighting conditions
  • Easy menu-driven operation with continuous diagnostic tests to ensure proper operation
  • Dual output of conductivity and temperature
  • Automatic temperature compensation
  • Nonvolatile memory retains settings during power outage
$1,150.00USD / Each
$178.00USD / Each
for an additional
$178.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Control TypeOn/Off
  • Display TypeLCD
  • Housing MaterialNEMA 4 (IP65)
  • Min Temperature (° C)-30
  • Max Temperature (° C)130
  • Min Temperature (° F)-22
  • Max Temperature (° F)266
  • Temperature CompensationAutomatic or Manual
  • Password ProtectionYes
  • Output Current4-20mA (x2)
  • Output Relay2 SPDT relays
  • DescriptionContacting Conductivity Transmitter/Controller, 1/4 DIN
Includes
mounting fittings.
Key Features
  • Large, graphical LCD with auto-sense backlight and contrast can be easily viewed in all lighting conditions
  • Easy menu-driven operation with continuous diagnostic tests to ensure proper operation
  • Dual output of conductivity and temperature
  • Automatic temperature compensation
  • Nonvolatile memory retains settings during power outage
  • Panel- or wall-mount NEMA 4X, IP65 rated housing
More About this Item
Contacting conductivity and resistivity transmitter/controller is designed for ultrapure water, sanitary, boiler/condensate, and general-purpose applications. The analyzer offers a wide range of useful features: two adjustable high/low relays, one wash control, and two isolated 4 to 20 mA outputs (one for the selected measurement and one for temperature), and enhanced measurement stability with sample averaging functionality. Automatic calibration with preset standards lets you enter in the sensor''s calibration constant to fully calibrate the analyzer. Calibration data can be password protected with a programmable security code.
ACCESSORIES
Call Us At1-847-549-7600
Live chat