Setra SRPM-R05WB-V1-E Room Pressure Monitor/Alarm, +/-0.05" WC, 120/240 VAC
Representative image only.
Setra Mfr # SRPM-R05WB-V1-E – Item # EW-98074-10
Designed for critical low differential pressure applications
  • Touch-screen display
  • Password enabled
  • Local audible alarm
  • Variable alarm delay
  • Bar graph display
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Accuracy±0.5% full-scale
  • Min Pressure (in WC)-0.05
  • Max Pressure (in WC)0.05
  • CaseFire-retardant plastic
  • Process Connection1/4\" hose barb
  • Unit Of Measurement\" WC
  • Display128 x 128 pixel, backlit LCD
  • Battery Type120/240 VAC
  • Electrical Connectionsterminal block
  • Output4 to 20 mA, 0 to 5 VDC, 0 to 10 VDC
  • Media CompatibilityAir and nonconductive, nonexplosive gases
  • DescriptionRoom Pressure Monitor/Alarm, +/-0.05\" WC, 120/240 VAC
  • Warranty1 year
1 Year WarrantyConformite Europeenne
Key Features
  • Touch-screen display
  • Password enabled
  • Local audible alarm
  • Variable alarm delay
  • Bar graph display
  • 1 year warranty
More About this Item
These room pressure monitors provide stringent pressure monitoring and alarming. The pressure monitor can be configured to monitor the positive or negative airflow in protected environments. Touch screen allows convenient fingertip access to menus that guide you through setup and operation.
Visual red and green LEDs and a local audible alarm to alert personnel to system status. The monitor also features a SPST relay output for remote indication of system status. True differential pressure is measured and displayed with 0.001 inches of water column resolution. Additional features include multiple selectable analog outputs, output and display averaging, and door status indication.
Applications - Pharmaceutical manufacturing, Semiconductor manufacturing, Cleanrooms, Research laboratories