Seward BA6141 Stomacher bags for Stomacher 400C; round bottom; 500/cs
Representative image only.
Seward Mfr # BA6141 – Item # EW-36850-93
  • Manufactured from high-grade, food quality polythene
  • Minimum 65 micron film thickness provides high-impact strength
  • All Seward Stomacher Bags are manufactured from virgin LDPE polymer. The only seal is at the base.
  • Compatible with all lab paddle blender, homogenizers.
  • All Seward Stomacher Bags are gamma irradiated at 25 to 50kGy to guarantee sterility.
PRODUCT OPTIONS
$157.00USD / Case of 500
SPECS & DESCRIPTION
Specifications & Description
  • Bag TypePaddle Blender Bags
  • Bag Capacity (mL)400
  • Bag Capacity (L)0.4
  • DescriptionStomacher bags for Stomacher 400C; round bottom; 500/cs
Key Features
  • Manufactured from high-grade, food quality polythene
  • Minimum 65 micron film thickness provides high-impact strength
  • All Seward Stomacher Bags are manufactured from virgin LDPE polymer. The only seal is at the base.
  • Compatible with all lab paddle blender, homogenizers.
  • All Seward Stomacher Bags are gamma irradiated at 25 to 50kGy to guarantee sterility.
More About this Item
Choose from a variety of bags to use with your paddle blender. Blending bags are made from nontoxic polyethylene. Bags with wire closures ensure samples stay in the bag and feature a convenient write-on strip.
The Stomacher Bag range has been designed to fulfil all applications from sample blending, straining and storage, including bags designed for extensive blending (processes requiring more than 15 minutes on the machine) and small samples for life science applications.
Our Stomacher bags are made from high quality food grade virgin LDPE granule, extruded to 65µm film thickness. All Stomacher blender bags are irradiated sterile. A certificate or irridiation is available upon request.
ACCESSORIES
Call Us At1-847-549-7600
Live chat