This item is now discontinued. We recommend the following as suggested replacements: EW-87701-04
Shimpo DT-725KIT-220V Digital Stroboscope Kit; 220 VAC
Representative image only.
Shimpo Mfr # DT-725KIT-230V – Item # EW-53560-32
Freezes rotating or reciprocating machinery for visual inspection without stopping production
  • Bright LED display aids viewing in poorly lit areas
  • External trigger allows unit to be automatically synchronized with equipment
  • Continuous duty cycle eliminates need to shut down for cooling
  • Phase shift allows visual analysis of rotating/reciprocating objects
  • Easy adjust flash rate controller dial
  • Synchronous output enables strobe to drive other strobes
  • Easy tri-pod or permanent mounting with integral mounting thread
  • Internal, fast-charge rechargeable battery supply
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Min Flash Range (fpm)40
  • Max Flash Range (fpm)12500
  • Flash Accuracy (fpm)±0.02% of reading
  • Display Type5 Digit LED
  • Battery TypeInternal rechargeable battery
  • DescriptionDigital Stroboscope kit, 220 VAC, W/Charger Case/Flash Tube and external triggering
  • Warranty1 Year
1 Year Warranty
Includes
Stroboscope, 230 VAC adapter/charger, spare flashtube, and protective carrying case.
Key Features
  • Bright LED display aids viewing in poorly lit areas
  • External trigger allows unit to be automatically synchronized with equipment
  • Continuous duty cycle eliminates need to shut down for cooling
  • Phase shift allows visual analysis of rotating/reciprocating objects
  • Easy adjust flash rate controller dial
  • Synchronous output enables strobe to drive other strobes
  • Easy tri-pod or permanent mounting with integral mounting thread
  • Internal, fast-charge rechargeable battery supply
  • 1 year warranty
More About this Item
These light weight stroboscopes are designed for quality control, motion analysis, vibration problems, maintenance work, experiments in engineering school projects, production line checks, examination and speed measurement of motors under various loads, gear movement and engagement, alignment of parts, cams, propellers, fan blades, centrifuges, spindles, turbines and other machinery exhibiting repetitive motion. They also provide pertinent information on the motion of vibrating parts, oil leaks, liquid spray patterns, and a host of other industrial and scientific applications.
The stroboscopes also feature phase shifting, rotary pulse indicator, floating decimal point and LED digital display, slave functions, and manual flash rate control. As electronic digital tachometers, you can measure RPM without contact or special markings.