Shimpo DT315AEB Digital Battery Powered Stroboscope
Representative image only.
Shimpo Mfr # DT-315AEB – Item # EW-53560-37
  • Easily carry external battery supply with shoulder harness
SPECS & DESCRIPTION
Specifications & Description
  • Min Flash Range (fpm)40
  • Max Flash Range (fpm)35000
  • Flash Accuracy (fpm)±0.01% of reading
  • Display Type5 Digit LED
  • Battery TypeExternal Rechargeable Battery
  • DescriptionDigital External Battery Powered Stroboscope with external triggering, 0 to 35,000 fpm
Key Features
  • Easily carry external battery supply with shoulder harness
More About this Item
The DT-315AEB Portable Stroboscope with rechargeable battery incorporates a robust, extruded aluminum housing with a precise, repeatable accuracy allowing it to be the solution for the most demanding applications. A truly portable unit, the DT-315AEB operates independent of line power via an exterior battery pack that can be carried with a convenient shoulder harness or set down to allow the operator full movement for visual inspection. When fully charged, rechargeable batteries power the strobe for approximately four continuous hours.