Shimpo FG-HK Hook Adapter for FG Series Force Gauges
Representative image only.
Shimpo Mfr # FG-HK – Item # EW-93957-21
Choose from a range of accessories to assist you
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionHook Adapter with M6 threading for FG Series Force Gauges
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
This replacement hook tip adapter comes standard with any FG-7000 series force gauge. It uses an M6 thread.
Please contact our Application Specialists for help in selecting an adapter tip for your testing needs.