Shimpo FGS-100PXH Motorized Force Gauge Test Stand, Horizontal standard
Representative image only.
Shimpo Mfr # FGS-100PXH – Item # EW-93950-94
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (lbs)50
  • Capacity (kg)22
  • Display4-digit, 0.375 in H LED
  • Power (VAC)120
  • DescriptionMotorized Force Gauge Test Stand, Horizontal standard
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
The motorized stands feature user programmable set points: speed, distance, and stop time. Single cycle and continuous cycling capabilities give you consistent testing parameters. Select high and low force set points through the RS-232 interface or when used with gauges with RS-232 output. Motorized stands have a four-digit, 0.41"H LED — read in inches or millimeters. Available in vertical or horizontal models to suit your application.