SHOWA 55-09 Liquid-Proof Latex Gloves, Rolled Cuff, large, 15"L, 12 pairs/pk
Representative image only.
SHOWA Mfr # 55-09 – Item # EW-86288-42
Superior 50 mil thickness
  • Ideal for sandblasting and electroplating
PRODUCT OPTIONS
$126.25USD / Pkg of 12
BUY MORE & SAVE
  • 1-5$126.25USD / Pkg of 12
  • 6+$113.63USD / Pkg of 12
SPECS & DESCRIPTION
Specifications & Description
  • MaterialLatex
  • Thickness (mil)50
  • SizeLarge
  • Length (in)15
  • Length (cm)38
  • ColorBlack and Red
  • SterileNo
  • DescriptionLiquid-Proof Latex Gloves, Rolled Cuff, large, 15\"L, 12 pairs/pk
Key Features
  • Ideal for sandblasting and electroplating
More About this Item
HD™ heavy-duty latex gloves feature two-color construction that shows wear—gives added safety for sandblasting and electroplating. Not designed for electrical use.