SHOWA 1005-Sm Latex Gloves, 5 mil, small
Representative image only.
SHOWA Mfr # 1005-Sm – Item # EW-86431-50
Ergonomically designed for better comfort, fit, and grip
  • Offers outstanding strength, durability, and wear
  • Ambidextrous
PRODUCT OPTIONS
$23.90USD / Box of 100
BUY MORE & SAVE
  • 1-9$23.90USD / Box of 100
  • 10+$21.51USD / Box of 100
SPECS & DESCRIPTION
Specifications & Description
  • MaterialLatex
  • Thickness (mil)5
  • SizeSmall
  • Length (in)9 1/2
  • Length (cm)24
  • ColorGreen
  • ComplianceFDA 21 CFR 170-199
  • SterileNo
  • DescriptionLatex Gloves, 5 mil, small
Key Features
  • Offers outstanding strength, durability, and wear
  • Ambidextrous
More About this Item
Derma Thin's specially formulated, natural-rubber compound features the outstanding strength, durability, wear and dexterity. Material is less susceptible to tearing or developing holes than similar-gauge vinyl gloves. Gloves are lightly powdered with USP-grade corn starch instead of talc for easy donning and doffing to reduce the chance of contamination.