Showa Best 7005PFXL Laboratory Grade Gloves, powder-free, X-large
Representative image only.
Showa Best Mfr # 7005PFXL – Item # EW-86287-83
Reduce hand fatigue by keeping your hands cool and dry
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
  • Nonlatex—made of soft 100% nitrile
  • Three times the puncture-resistance of natural rubber or vinyl
  • Superior tactile-sensitivity, comfort, and dexterity
  • Lightweight and disposable
PRODUCT OPTIONS
$21.50USD / Pkg of 100
BUY MORE & SAVE
  • 1-9$21.50USD / Pkg of 100
  • 10+$19.35USD / Pkg of 100
SPECS & DESCRIPTION
Specifications & Description
  • MaterialNitrile
  • Thickness (mil)4
  • SizeX-Large
  • Length (in)9 1/2
  • Length (cm)24
  • ColorLight Blue
  • ComplianceFDA 21 CFR 170-199
  • SterileNo
  • PowderedNo
  • DescriptionLaboratory Grade Gloves, powder-free, X-large
Key Features
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
  • Nonlatex—made of soft 100% nitrile
  • Three times the puncture-resistance of natural rubber or vinyl
  • Superior tactile-sensitivity, comfort, and dexterity
  • Lightweight and disposable
More About this Item
N-DEX® gloves are made from a special low-modulus, nonlatex, soft nitrile formulation. They offer greater tactile-sensitivity, comfort, dexterity, and puncture-resistance than latex gloves. All N-DEX® nitrile gloves are ambidextrous and blue in color.
General and Laboratory Grade N-DEX® Gloves are ideal for use anywhere you need to protect your hands from the environment. Box of 100.