SHOWA N-DEX Industrial Grade Nitrile Gloves, Size XL; 50/PK
Representative image only.
SHOWA Mfr # 8005XL – Item # EW-86287-63
Reduce hand fatigue by keeping your hands cool and dry
  • Nonlatex—made of soft 100% nitrile
  • Three times the puncture-resistance of natural rubber or vinyl
  • Superior tactile-sensitivity, comfort, and dexterity
  • Lightweight and disposable
PRODUCT OPTIONS
$25.00USD / Pkg of 50
BUY MORE & SAVE
  • 1-9$25.00USD / Pkg of 50
  • 10+$22.50USD / Pkg of 50
SPECS & DESCRIPTION
Specifications & Description
  • MaterialNitrile
  • Thickness (mil)8
  • SizeX-Large
  • Length (in)9 1/2
  • Length (cm)24
  • ColorWhite
  • ComplianceFDA 21 CFR 170-199
  • SterileNo
  • DescriptionNon-latex Industrial Gloves; size, extra-large; 50/pack
Conformite Europeenne
Key Features
  • Nonlatex—made of soft 100% nitrile
  • Three times the puncture-resistance of natural rubber or vinyl
  • Superior tactile-sensitivity, comfort, and dexterity
  • Lightweight and disposable
More About this Item
N-DEX® gloves are made from a special low-modulus, nonlatex, soft nitrile formulation. They offer greater tactile-sensitivity, comfort, dexterity, and puncture-resistance than latex gloves. All N-DEX® nitrile gloves are ambidextrous and blue in color.
Industrial Grade N-DEX® Plus Gloves are thicker for greater puncture and tear resistance. Gloves provide good resistance to chemicals and abrasion. Ideal for tough lab conditions, assembly, or manufacturing. Available only in low-powder versions. Box of 50.