Side-Inlet Sanitary Strainer Core; 1/4" Perforations
Representative image only.
Item # EW-29590-72
Protect products and expensive process equipment from coarse or fine particulates
  • 316 SS fluid contact surfaces are polished to a smooth, blemish free finish
  • Standard clamp sizes allow easy installation or retrofit
  • Side-inlet design lets you clean housing and change screens without disconnecting the strainer from your process line
PRODUCT OPTIONS
$470.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Filter TypeStrainer
  • DescriptionSide-Inlet Sanitary Strainer Core; 1/4" Perforations
Includes
strainer assemblies include housing, perforated core, internal cap and spring, assembly clamp and gasket. Order wire mesh overscreens of the desired mesh size separately.
Key Features
  • 316 SS fluid contact surfaces are polished to a smooth, blemish free finish
  • Standard clamp sizes allow easy installation or retrofit
  • Side-inlet design lets you clean housing and change screens without disconnecting the strainer from your process line
More About this Item
Sanitary strainers are ideal for particulate removal or pre-filtration in the processing of foods, beverages, pharmaceuticals, cosmetics, and other critical compounds. Housings are available in in-line and side-inlet configurations.