Simpson 1227A 06215 Dc Milliammeter 0-20 2 1/2in Model:06215
Representative image only.
Simpson Mfr # 1227A | 06215 – Item # EW-68488-70
SPECS & DESCRIPTION
Specifications & Description
  • Control Input mA0-20
  • Display Type2.5\" analog dial
  • DescriptionDc Milliammeter 0-20 2 1/2in Model:06215
More About this Item
FEATURES
  • Clear acrylic window for wide angle visibility
  • Rugged black phenolic case
  • Black spade pointer for easy distant reading
  • UL recognized
Additional ranges available. Contact Davis