Simpson 1227A 09560 Voltmeter, 0-10 VDC, 2.5 UL WV
Representative image only.
Simpson Mfr # 09560 – Item # EW-68488-88
SPECS & DESCRIPTION
Specifications & Description
  • Display Type2.5\" analog dial
  • Max DC Voltage VDC (VDC)10
  • Min DC Voltage VDC (VDC)0
  • DescriptionVoltmeter, 0-10 VDC, 2.5 UL WV
More About this Item
FEATURES
  • Clear acrylic window for wide angle visibility
  • Rugged black phenolic case
  • Black spade pointer for easy distant reading
  • UL recognized
Additional ranges available. Contact Davis