Simpson 1257MD 02609 Ac Ammeter W/Iron Vane 0-25 2 1/2in Model:02609
Representative image only.
Simpson Mfr # 02609 – Item # EW-68487-56
SPECS & DESCRIPTION
Specifications & Description
  • Display Type2.5\" analog dial
  • Max DC Current mA (mA)25000
  • Min DC Current mA (mA)0
  • DescriptionAc Ammeter W/Iron Vane 0-25 2 1/2in Model:02609
More About this Item
FEATURES
  • Clear acrylic window for wide angle visibility
  • Rugged black phenolic case
  • Black spade pointer for easy distant reading
  • UL recognized
Additional ranges available. Contact Davis