Simpson 1327 04420 Dc Microammeter 25-0-25 3 1/2in Model:04420
Representative image only.
Simpson Mfr # 1327 | 04420 – Item # EW-68488-59
SPECS & DESCRIPTION
Specifications & Description
  • Display Type3.5\" analog dial
  • Max DC Current mA (mA)250000000
  • Min DC Current mA (mA)0
  • DescriptionDc Microammeter 25-0-25 3 1/2in Model:04420
More About this Item
FEATURES
  • Clear acrylic window for wide angle visibility
  • Rugged black phenolic case
  • Black spade pointer for easy distant reading
  • UL recognized
Additional ranges available. Contact Davis