Simpson 1327A 02730 Ammeter, 0-50 A DC, 3.5 UL WV
Representative image only.
Simpson Mfr # 02730 – Item # EW-68488-42
SPECS & DESCRIPTION
Specifications & Description
  • Display Type3.5\" analog dial
  • Max DC Current mA (mA)50000
  • Min DC Current mA (mA)0
  • DescriptionAmmeter, 0-50 A DC, 3.5 UL WV
More About this Item
FEATURES
  • Clear acrylic window for wide angle visibility
  • Rugged black phenolic case
  • Black spade pointer for easy distant reading
  • UL recognized
Additional ranges available. Contact Davis