Simpson 1327A 06390 Dc Milliammeter 0-100 3 1/2in Model:06390
Representative image only.
Simpson Mfr # 06390 – Item # EW-68488-64
$143.75USD / Each
$112.25 - $375.00USD / Each
for an additional
$112.25USD / Each
for an additional
$133.00USD / Each
for an additional
$206.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Display Type3.5\" analog dial
  • Max DC Current mA (mA)100
  • Min DC Current mA (mA)0
  • DescriptionDc Milliammeter 0-100 3 1/2in Model:06390
More About this Item
FEATURES
  • Clear acrylic window for wide angle visibility
  • Rugged black phenolic case
  • Black spade pointer for easy distant reading
  • UL recognized
Additional ranges available. Contact Davis