Simpson 1359M 35077 4.5in Ac Ammeters W/ Iron Vane Movement
Representative image only.
Simpson Mfr # 1359M | 35077 – Item # EW-68487-23
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Display Type4.5\" analog dial
  • Max DC Current mA (mA)10000
  • Min DC Current mA (mA)0
  • Description4.5in Ac Ammeters W/ Iron Vane Movement
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
FEATURES
  • Clear acrylic window for wide angle visibility
  • Rugged black phenolic case
  • Black spade pointer for easy distant reading
  • UL recognized
Additional ranges available. Contact Davis